accessibility menu, dialog, popup

UserName

CAS RN 30007-47-7

IUPAC Name: 5-bromo-5-nitro-1,3-dioxane
Synonyms: 5-bromo-5-nitro-1,3-dioxane bronidox
Molecular Weight (g/mol): 212
Molecular Formula: C4H6BrNO4
InChi Key: XVBRCOKDZVQYAY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1(Br)COCOC1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)