missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-5-nitro-1,3-dioxane, 98%

Catalog No. p-7043082
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6544306 5 g
AAJ6544314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6544306 Supplier Thermo Scientific Chemicals Supplier No. J6544306
Only null left

5-Bromo-5-nitro-1,3-dioxane, also known as bronidox, shows antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi. | CAS: 30007-47-7 | C4H6BrNO4 | 211.999 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• 5-Bromo-5-nitro-1,3-dioxane is a chemical compound showing antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi
• This compound is capable of promoting the oxidation of essential protein thiols, causing inhibition of enzyme activity leading to the inhibition of microbial growth

Application

• 5-Bromo-5-nitro-1,3-dioxane has been used as a stabilizer and preserving agent for biological molecules and solutions, including antibodies and antisera
• It can be used alone or combined with methylisothiazolinone, which is also considered to be an effective preservative

Chemical Identifiers

CAS 30007-47-7
Molecular Formula C4H6BrNO4
Molecular Weight (g/mol) 211.999
MDL Number MFCD00101855
InChI Key XVBRCOKDZVQYAY-UHFFFAOYSA-N
Synonym bronidox, 1,3-dioxane, 5-bromo-5-nitro, 5-bromo-5-nitro-m-dioxane, unii-u184i9qbnm, 5-brom-5-nitro-1,3-dioxan, m-dioxane, 5-bromo-5-nitro, 5-brom-5-nitro-1,3-dioxan german, u184i9qbnm, bronidox l, acmc-209hcf
PubChem CID 1807
IUPAC Name 5-bromo-5-nitro-1,3-dioxane
SMILES C1C(COCO1)([N+](=O)[O-])Br

Specifications

Melting Point 58°C to 61°C
Color White
Quantity 5 g
Solubility Information Soluble in water at 12.5mg/ml
Formula Weight 212
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromo-5-nitro-1,3-dioxane
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 250-001-7
RTECSNumber JG9650000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only