missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-chloro-3-indolyl beta-D-Glucuronide Sodium Salt 98.0+%, TCI America™
$94.93 - $484.15
Chemical Identifiers
| CAS | 129541-41-9 |
|---|---|
| Molecular Formula | C14H12BrClNNaO7 |
| Molecular Weight (g/mol) | 444.594 |
| MDL Number | MFCD00135782 |
| InChI Key | IBLSVGDGSKUDCT-ILIJQVQCSA-M |
| Synonym | x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt |
| PubChem CID | 53384407 |
| IUPAC Name | sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate |
| SMILES | C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
B362110MG
|
N/A | 10 mg |
Each for $94.93
|
|
|||||
|
B3621100MG
|
N/A | 100 mg |
Each for $484.15
|
|
|||||
Chemical Identifiers
| 129541-41-9 | |
| 444.594 | |
| IBLSVGDGSKUDCT-ILIJQVQCSA-M | |
| 53384407 | |
| C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+] |
| C14H12BrClNNaO7 | |
| MFCD00135782 | |
| x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt | |
| sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate |
Specifications
| 129541-41-9 | |
| C14H12BrClNNaO7 | |
| x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt | |
| C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+] | |
| 444.594 | |
| 444.59 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00135782 | |
| IBLSVGDGSKUDCT-ILIJQVQCSA-M | |
| sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate | |
| 53384407 | |
| ≥98.0% (HPLC,T) | |
| 5-Bromo-4-chloro-3-indolyl beta-D-Glucuronide Sodium Salt [for Biochemical Research] |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator