missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl beta-D-Glucuronide Sodium Salt 98.0+%, TCI America™

Supplier:  TCI America B3621100MG

Quantity 100 mg

 View more versions of this product

Catalog No. B3621100MG


Only null left

Description

Description

Specifications

Chemical Identifiers

129541-41-9
444.594
IBLSVGDGSKUDCT-ILIJQVQCSA-M
53384407
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+]
C14H12BrClNNaO7
MFCD00135782
x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt
sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate

Specifications

5-Bromo-4-chloro-3-indolyl beta-D-Glucuronide Sodium Salt [for Biochemical Research]
White-Yellow
100 mg
x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+]
444.594
444.59
Crystalline Powder
129541-41-9
C14H12BrClNNaO7
MFCD00135782
IBLSVGDGSKUDCT-ILIJQVQCSA-M
sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
53384407
≥98.0% (HPLC,T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions