missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt hydrate, 98%

CAS: 114162-64-0 | C20H26BrClN2O7 | 521.79 g/mol

$316.39 - $316.39

Chemical Identifiers

CAS 114162-64-0
Molecular Formula C20H26BrClN2O7
Molecular Weight (g/mol) 521.79
MDL Number MFCD13185253
InChI Key JXCKZXHCJOVIAV-VLQIOUPANA-N
Synonym 5-bromo-4-chloro-3-indolyl, a-d-glucuronide cyclohe, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
PubChem CID 126842357
IUPAC Name (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium;hydrate
SMILES NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC331190250
SDS View Documents
Thermo Scientific Chemicals
331190250
25 mg Glass bottle
Each for $316.39
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

114162-64-0
521.79
JXCKZXHCJOVIAV-VLQIOUPANA-N
126842357
NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O
C20H26BrClN2O7
MFCD13185253
5-bromo-4-chloro-3-indolyl, a-d-glucuronide cyclohe, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium;hydrate

Specifications

114162-64-0
100.0
Colorless or White
97.5% min. (HPLC)
C20H26BrClN2O7
25 mg
Solubility in water: soluble
NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O
521.79
521.79
Crystals, Powder or Needles
97.5
230°C
Authentic
Glass bottle
MFCD13185253
5-bromo-4-chloro-3-indolyl, a-d-glucuronide cyclohe, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
JXCKZXHCJOVIAV-VLQIOUPANA-N
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium;hydrate
126842357
98%
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt hydrate, 98%
Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

LIGHT SENSITIVE

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only