accessibility menu, dialog, popup

UserName

CAS RN 114162-64-0

IUPAC Name: (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid; cyclohexanamine
Synonyms: X-gluc; cyclohexylamine
Molecular Weight (g/mol): 521.79
Molecular Formula: C20H26BrClN2O7
InChi Key: JXCKZXHCJOVIAV-VLQIOUPANA-N
SMILES: NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)