missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl beta-D-Glucuronide Cyclohexylammonium Salt 98.0+%, TCI America™

Catalog No. p-7123053
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B362010MG 10 mg
B3620100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B362010MG Supplier TCI America Supplier No. B362010MG

Chemical Identifiers

CAS 114162-64-0
Molecular Formula C20H26BrClN2O7
Molecular Weight (g/mol) 521.79
MDL Number MFCD13185253
InChI Key JXCKZXHCJOVIAV-VLQIOUPANA-N
Synonym 5-bromo-4-chloro-3-indolyl, a-d-glucuronide cyclohe, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
PubChem CID 126842357
IUPAC Name (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid; cyclohexanamine
SMILES NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O

Specifications

Color White-Yellow
Quantity 10 mg
Formula Weight 521.79
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl beta-D-Glucuronide Cyclohexylammonium Salt [for Biochemical Research]
TSCA Yes
Recommended Storage Refrigerator