missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-iodo-2-hydroxypyridine, 97%

CAS: 381233-75-6 | C5H3BrINO | 299.893 g/mol

Supplier:  Thermo Scientific Chemicals H3349506

 View more versions of this product

Catalog No. AAH3349506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Bromo-3-iodo-2-hydroxypyridine
226°C to 227°C
MFCD08688586
Light sensitive
WAUPPDAXWKLNNU-UHFFFAOYSA-N
5-bromo-3-iodo-1H-pyridin-2-one
12027787
97%
381233-75-6
C5H3BrINO
5 g
5-bromo-3-iodopyridin-2 1h-one, 5-bromo-3-iodopyridin-2-ol, 5-bromo-3-iodo-pyridin-2-ol, 5-bromo-2-hydroxy-3-iodopyridine, 5-bromo-3-iodo-1,2-dihydropyridin-2-one, 2-hydroxy-3-iodo-5-bromopyridine, 2 1h-pyridinone,5-bromo-3-iodo, 2 1h-pyridinone, 5-bromo-3-iodo, 5-bromo-3-iodo-2-hydroxypyridine, acmc-1cu7j
C1=C(C(=O)NC=C1Br)I
299.893
299.89

Chemical Identifiers

381233-75-6
299.893
WAUPPDAXWKLNNU-UHFFFAOYSA-N
5-bromo-3-iodo-1H-pyridin-2-one
C5H3BrINO
MFCD08688586
12027787
C1=C(C(=O)NC=C1Br)I
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only