missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-iodo-2-hydroxypyridine, 97%

Catalog No. p-7032140
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3349503 1 g
AAH3349506 5 g
AAH3349514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3349503 Supplier Thermo Scientific Chemicals Supplier No. H3349503
Only null left

CAS: 381233-75-6 | C5H3BrINO | 299.893 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 381233-75-6
Molecular Formula C5H3BrINO
Molecular Weight (g/mol) 299.893
MDL Number MFCD08688586
InChI Key WAUPPDAXWKLNNU-UHFFFAOYSA-N
PubChem CID 12027787
IUPAC Name 5-bromo-3-iodo-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Br)I

Specifications

Melting Point 226°C to 227°C
Quantity 1 g
Sensitivity Light sensitive
Synonym 5-bromo-3-iodopyridin-2 1h-one, 5-bromo-3-iodopyridin-2-ol, 5-bromo-3-iodo-pyridin-2-ol, 5-bromo-2-hydroxy-3-iodopyridine, 5-bromo-3-iodo-1,2-dihydropyridin-2-one, 2-hydroxy-3-iodo-5-bromopyridine, 2 1h-pyridinone,5-bromo-3-iodo, 2 1h-pyridinone, 5-bromo-3-iodo, 5-bromo-3-iodo-2-hydroxypyridine, acmc-1cu7j
Formula Weight 299.89
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-iodo-2-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only