missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-thiophenemethanol 97.0+%, TCI America™
Supplier: TCI America B46515G
Specifications
| 5-Bromo-2-thiophenemethanol | |
| Yellow | |
| MFCD06202626 | |
| 5-bromothien-2-yl methanol, 5-bromothiophen-2-yl methanol, 5-bromo-2-thiophenemethanol, 2-thiophenemethanol, 5-bromo, 5-bromothiophene-2-methanol, 5-bromo-2-thienyl methanol, ksc646o4p, 2-thiophenemethanol,5-bromo, 5-bromo-2-thiophenyl methanol, 5-bromo-2-hydroxymethylthiophene | |
| OCC1=CC=C(Br)S1 | |
| 193.06 | |
| 193.06 | |
| Liquid |
| 79387-71-6 | |
| C5H5BrOS | |
| 5 g | |
| DBRSLMCLMMFHEF-UHFFFAOYSA-N | |
| (5-bromothiophen-2-yl)methanol | |
| 11435467 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 79387-71-6 | |
| 193.06 | |
| DBRSLMCLMMFHEF-UHFFFAOYSA-N | |
| (5-bromothiophen-2-yl)methanol |
| C5H5BrOS | |
| MFCD06202626 | |
| 11435467 | |
| OCC1=CC=C(Br)S1 |
Safety and Handling
TSCA : No