missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-thiophenemethanol 97.0+%, TCI America™

Catalog No. p-7123669
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B46511G 1 g
B46515G 5 g
2 options

Catalog No. B46511G

Supplier: TCI America B46511G

Only null left

Chemical Identifiers

CAS 79387-71-6
Molecular Formula C5H5BrOS
Molecular Weight (g/mol) 193.06
MDL Number MFCD06202626
InChI Key DBRSLMCLMMFHEF-UHFFFAOYSA-N
PubChem CID 11435467
IUPAC Name (5-bromothiophen-2-yl)methanol
SMILES OCC1=CC=C(Br)S1

Specifications

Color Yellow
Quantity 1 g
Synonym 5-bromothien-2-yl methanol, 5-bromothiophen-2-yl methanol, 5-bromo-2-thiophenemethanol, 2-thiophenemethanol, 5-bromo, 5-bromothiophene-2-methanol, 5-bromo-2-thienyl methanol, ksc646o4p, 2-thiophenemethanol,5-bromo, 5-bromo-2-thiophenyl methanol, 5-bromo-2-hydroxymethylthiophene
Formula Weight 193.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-thiophenemethanol