missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-thiophenecarboxylic Acid 98.0+%, TCI America™

Catalog No. B23025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23025G 5 g
B23021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23025G Supplier TCI America Supplier No. B23025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7311-63-9
Molecular Formula C5H2BrO2S
Molecular Weight (g/mol) 206.03
MDL Number MFCD00079725
InChI Key COWZPSUDTMGBAT-UHFFFAOYSA-M
Synonym 5-bromo-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-bromo, 2-bromo-5-carboxythiophene, 2-bromothiophene-5-carboxylic acid, 5-bromo-thiophene-2-carboxylic acid, 5-bromothiophene-2-carboxylicacid, zlchem 973, pubchem7368, acmc-209opm, maybridge1_002056
PubChem CID 349115
IUPAC Name 5-bromothiophene-2-carboxylate
SMILES [O-]C(=O)C1=CC=C(Br)S1

Specifications

Melting Point 142°C
Color Yellow
Quantity 5 g
Formula Weight 207.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-thiophenecarboxylic Acid