missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-thiophenecarboxaldehyde, 97%

CAS: 4701-17-1 | C5H3BrOS | 191.04 g/mol

$116.43 - $116.43

Chemical Identifiers

CAS 4701-17-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005432
InChI Key GFBVUFQNHLUCPX-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde
PubChem CID 78428
IUPAC Name 5-bromothiophene-2-carbaldehyde
SMILES BrC1=CC=C(S1)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC166650050
SDS View Documents
Thermo Scientific Chemicals
166650050
5 mL Glass bottle
Each for $116.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4701-17-1
191.04
GFBVUFQNHLUCPX-UHFFFAOYSA-N
78428
BrC1=CC=C(S1)C=O
C5H3BrOS
MFCD00005432
5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde
5-bromothiophene-2-carbaldehyde

Specifications

4701-17-1
100.0
1.6100g/mL
Authentic
Glass bottle
1.6380 to 1.641
5 mL
1.61
GFBVUFQNHLUCPX-UHFFFAOYSA-N
5-bromothiophene-2-carbaldehyde
78428
97%
5-Bromo-2-thiophenecarboxaldehyde
96.0
Yellow to Brown
98°C
96% min. (GC)
C5H3BrOS
MFCD00005432
17, I, 148
5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde
BrC1=CC=C(S1)C=O
191.04
191.05
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-176-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only