missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-phenylpyridine 98.0+%, TCI America™

Catalog No. p-7123901
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50181G 1 g
B50185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50181G Supplier TCI America Supplier No. B50181G

Chemical Identifiers

CAS 27012-25-5
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD04114181
InChI Key PRNGIODVYLTUKH-UHFFFAOYSA-N
Synonym 2-phenyl-5-bromopyridine, 3-bromo-6-phenylpyridine, pyridine, 5-bromo-2-phenyl, 5-bromo-2-phenyl-pyridine, pubchem21997, acmc-20am5l, 5-bromo-2-phenylpyridine, abbypharma ap-11-5994
PubChem CID 2762862
IUPAC Name 5-bromo-2-phenylpyridine
SMILES C1=CC=C(C=C1)C2=NC=C(C=C2)Br

Specifications

Melting Point 75°C
Color White
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-phenylpyridine