accessibility menu, dialog, popup

UserName

NBr

CAS RN 27012-25-5

IUPAC Name: 5-bromo-2-phenylpyridine
Synonyms: 5-bromo-2-phenylpyridine
Molecular Weight (g/mol): 234.1
Molecular Formula: C11H8BrN
InChi Key: PRNGIODVYLTUKH-UHFFFAOYSA-N
SMILES: BrC1=CN=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results