missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-phenyl-7-azaindole, 97%, Thermo Scientific Chemicals

$561.37 - $561.37

Chemical Identifiers

CAS 953414-75-0
Molecular Formula C13H9BrN2
Molecular Weight (g/mol) 273.13
MDL Number MFCD11109822
InChI Key QLKZJMXSXCDCCF-UHFFFAOYSA-N
Synonym 5-bromo-2-phenyl-1h-pyrrolo 2,3-b pyridine, 5-bromo-2-phenyl-7-azaindole, 1h-pyrrolo 2,3-b pyridine,5-bromo-2-phenyl, 2-phenyl-5-bromo-1h-pyrrolo 2,3-b pyridine
PubChem CID 23643607
IUPAC Name 5-bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine
SMILES BrC1=CN=C2NC(=CC2=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3519303
View Documents
Thermo Scientific Chemicals
H3519303
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

953414-75-0
273.13
QLKZJMXSXCDCCF-UHFFFAOYSA-N
23643607
BrC1=CN=C2NC(=CC2=C1)C1=CC=CC=C1
C13H9BrN2
MFCD11109822
5-bromo-2-phenyl-1h-pyrrolo 2,3-b pyridine, 5-bromo-2-phenyl-7-azaindole, 1h-pyrrolo 2,3-b pyridine,5-bromo-2-phenyl, 2-phenyl-5-bromo-1h-pyrrolo 2,3-b pyridine
5-bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine

Specifications

953414-75-0
Brown
MFCD11109822
Light sensitive
QLKZJMXSXCDCCF-UHFFFAOYSA-N
5-bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine
23643607
97%
5-Bromo-2-phenyl-7-azaindole
287°C to 289°C
C13H9BrN2
1 g
5-bromo-2-phenyl-1h-pyrrolo 2,3-b pyridine, 5-bromo-2-phenyl-7-azaindole, 1h-pyrrolo 2,3-b pyridine,5-bromo-2-phenyl, 2-phenyl-5-bromo-1h-pyrrolo 2,3-b pyridine
BrC1=CN=C2NC(=CC2=C1)C1=CC=CC=C1
273.13
273.12
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only