accessibility menu, dialog, popup

UserName

CAS RN 953414-75-0

IUPAC Name: 5-bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine
Synonyms: 5-bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 273.13
Molecular Formula: C13H9BrN2
InChi Key: QLKZJMXSXCDCCF-UHFFFAOYSA-N
SMILES: BrC1=CN=C2NC(=CC2=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results