missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-nitrobenzotrifluoride 98.0+%, TCI America™
$82.79 - $238.72
Chemical Identifiers
| CAS | 344-38-7 |
|---|---|
| Molecular Formula | C7H3BrF3NO2 |
| Molecular Weight (g/mol) | 270.005 |
| MDL Number | MFCD00014706 |
| InChI Key | ZHLYHEDQTJZYFI-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol |
| PubChem CID | 136171 |
| IUPAC Name | 4-bromo-1-nitro-2-(trifluoromethyl)benzene |
| SMILES | C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-] |
Chemical Identifiers
| 344-38-7 | |
| 270.005 | |
| ZHLYHEDQTJZYFI-UHFFFAOYSA-N | |
| 136171 | |
| C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-] |
| C7H3BrF3NO2 | |
| MFCD00014706 | |
| 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol | |
| 4-bromo-1-nitro-2-(trifluoromethyl)benzene |
Specifications
| 344-38-7 | |
| Yellow | |
| C7H3BrF3NO2 | |
| 5 g | |
| ZHLYHEDQTJZYFI-UHFFFAOYSA-N | |
| 4-bromo-1-nitro-2-(trifluoromethyl)benzene | |
| 136171 | |
| ≥98.0% (GC) | |
| 5-Bromo-2-nitrobenzotrifluoride |
| 35°C | |
| 100°C | |
| MFCD00014706 | |
| 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol | |
| C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-] | |
| 270.005 | |
| 270.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : No