missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. B28535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28535G 5 g
B285325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28535G Supplier TCI America Supplier No. B28535G
Only null left

Chemical Identifiers

CAS 344-38-7
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.005
MDL Number MFCD00014706
InChI Key ZHLYHEDQTJZYFI-UHFFFAOYSA-N
Synonym 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol
PubChem CID 136171
IUPAC Name 4-bromo-1-nitro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 35°C
Color Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 270.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-nitrobenzotrifluoride