Learn More
5-Bromo-2-nitrobenzotrifluoride, 98+%
CAS: 344-38-7 | C7H3BrF3NO2 | 270.005 g/mol
$34.59 - $328.34
Chemical Identifiers
| CAS | 344-38-7 |
|---|---|
| Molecular Formula | C7H3BrF3NO2 |
| Molecular Weight (g/mol) | 270.005 |
| MDL Number | MFCD00014706 |
| InChI Key | ZHLYHEDQTJZYFI-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol |
| PubChem CID | 136171 |
| IUPAC Name | 4-bromo-1-nitro-2-(trifluoromethyl)benzene |
| SMILES | C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1763406
|
Thermo Scientific Chemicals
A1763406 |
5 g |
Each for $34.59
|
|
|||||
|
AAA1763414
|
Thermo Scientific Chemicals
A1763414 |
25 g |
Each for $115.99
|
|
|||||
|
AAA1763422
|
Thermo Scientific Chemicals
A1763422 |
100 g |
Each for $328.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 344-38-7 | |
| 270.005 | |
| ZHLYHEDQTJZYFI-UHFFFAOYSA-N | |
| 136171 | |
| C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-] |
| C7H3BrF3NO2 | |
| MFCD00014706 | |
| 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol | |
| 4-bromo-1-nitro-2-(trifluoromethyl)benzene |
Specifications
| 344-38-7 | |
| 1.799 | |
| >110°C (230°F) | |
| 1.518 | |
| 5 g | |
| 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol | |
| C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-] | |
| 270.005 | |
| 270.01 | |
| 5-Bromo-2-nitrobenzotrifluoride |
| 32°C to 35°C | |
| 95°C to 96°C (5 mmHg) | |
| C7H3BrF3NO2 | |
| MFCD00014706 | |
| 2650701 | |
| ZHLYHEDQTJZYFI-UHFFFAOYSA-N | |
| 4-bromo-1-nitro-2-(trifluoromethyl)benzene | |
| 136171 | |
| ≥98% |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only