missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-nitrobenzotrifluoride, 98+%

Catalog No. AAA1763422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1763422 100 g
AAA1763406 5 g
AAA1763414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1763422 Supplier Thermo Scientific Chemicals Supplier No. A1763422
Only null left

CAS: 344-38-7 | C7H3BrF3NO2 | 270.005 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 344-38-7
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.005
MDL Number MFCD00014706
InChI Key ZHLYHEDQTJZYFI-UHFFFAOYSA-N
Synonym 5-bromo-2-nitrobenzotrifluoride, 4-bromo-1-nitro-2-trifluoromethyl benzene, 2-nitro-5-bromobenzotrifluoride, 5-bromo-2-nitro-a,a,a-trifluorotoluene, 5-bromo-a-a,a-trifluoro-2-nitrotoluene, 4-bromo-1-nitro-2 trifluoromethyl benzene, 4-bromo-1-nitro-2-trifluoromethyl-benzene, benzene, 4-bromo-1-nitro-2-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-2-nitrotoluene, 2-nitro-5-bromotrifluorotoluol
PubChem CID 136171
IUPAC Name 4-bromo-1-nitro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 32°C to 35°C
Density 1.799
Boiling Point 95°C to 96°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.518
Quantity 100 g
Beilstein 2650701
Formula Weight 270.01
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-nitrobenzotrifluoride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only