missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylpyridine 98.0+%, TCI America™

Catalog No. p-7119690
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B25721G 1 g
B25725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B25721G Supplier TCI America Supplier No. B25721G

Chemical Identifiers

CAS 3430-13-5
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00661170
InChI Key OFKWIQJLYCKDNY-UHFFFAOYSA-N
Synonym 5-bromo-2-picoline, 2-methyl-5-bromopyridine, 5-bromo-2-methyl-pyridine, 3-bromo-6-methylpyridine, pyridine, 5-bromo-2-methyl, 5-bromopicoline, 5-bromo-2-methyl pyridine, 3-bromo-6-picoline, pubchem4058
PubChem CID 1201020
IUPAC Name 5-bromo-2-methylpyridine
SMILES CC1=CC=C(Br)C=N1

Specifications

Melting Point 36°C
Boiling Point 74°C
Quantity 1 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methylpyridine