accessibility menu, dialog, popup

UserName

CH3BrN

CAS RN 3430-13-5

IUPAC Name: 5-bromo-2-methylpyridine
Synonyms: 5-bromo-2-methylpyridine
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: OFKWIQJLYCKDNY-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results