missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylindole, 96%

Catalog No. p-4258811
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437560010 1 g Glass bottle
AC437560050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AC437560010 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 437560010
Only null left

CAS: 1075-34-9 | C9H8BrN | 210.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1075-34-9
Molecular Formula C9H8BrN
Molecular Weight (g/mol) 210.07
InChI Key BJUZAZKEDCDGRW-UHFFFAOYSA-N
PubChem CID 5003968
IUPAC Name 5-bromo-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 104.0°C to 106.0°C
Color Tan to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 5-bromo-2-methylindole, 2-methyl-5-bromoindole, 1h-indole, 5-bromo-2-methyl, 5-bromo-2-methylindol, pubchem9382, acmc-209xly, 5-bromo-2-methyl-indole, 2-methyl-5-bromo-1h-indole, 1h-indole,5-bromo-2-methyl
Formula Weight 210.07
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromo-2-methylindole
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
W

RUO – Research Use Only