accessibility menu, dialog, popup

UserName

CAS RN 1075-34-9

IUPAC Name: 5-bromo-2-methyl-1H-indole
Synonyms: 5-bromo-2-methyl-1H-indole
Molecular Weight (g/mol): 210.07
Molecular Formula: C9H8BrN
InChi Key: BJUZAZKEDCDGRW-UHFFFAOYSA-N
SMILES: CC1=CC2=CC(Br)=CC=C2N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)