missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylindole, 96%

Catalog No. AAH6684806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6684806 5 g
AAH6684803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6684806 Supplier Thermo Scientific Chemicals Supplier No. H6684806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1075-34-9 | C9H8BrN | 210.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1075-34-9
Molecular Formula C9H8BrN
Molecular Weight (g/mol) 210.074
MDL Number MFCD01863677
InChI Key BJUZAZKEDCDGRW-UHFFFAOYSA-N
PubChem CID 5003968
IUPAC Name 5-bromo-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)Br

Specifications

Melting Point 104°C to 107°C
Color Brown to White
Quantity 5 g
Beilstein 116647
Synonym 5-bromo-2-methylindole, 2-methyl-5-bromoindole, 1h-indole, 5-bromo-2-methyl, 5-bromo-2-methylindol, pubchem9382, acmc-209xly, 5-bromo-2-methyl-indole, 2-methyl-5-bromo-1h-indole, 1h-indole,5-bromo-2-methyl
Formula Weight 210.07
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 5-Bromo-2-methylindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only