missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-methylbenzaldehyde 95.0+%, TCI America™
$152.86 - $603.95
Chemical Identifiers
| CAS | 90050-59-2 |
|---|---|
| Molecular Formula | C8H7BrO |
| Molecular Weight (g/mol) | 199.047 |
| MDL Number | MFCD05662390 |
| InChI Key | YILPAIKZHXATHY-UHFFFAOYSA-N |
| PubChem CID | 10420289 |
| IUPAC Name | 5-bromo-2-methylbenzaldehyde |
| SMILES | CC1=C(C=C(C=C1)Br)C=O |
Chemical Identifiers
| 90050-59-2 | |
| 199.047 | |
| YILPAIKZHXATHY-UHFFFAOYSA-N | |
| 5-bromo-2-methylbenzaldehyde |
| C8H7BrO | |
| MFCD05662390 | |
| 10420289 | |
| CC1=C(C=C(C=C1)Br)C=O |
Specifications
| 90050-59-2 | |
| White-Yellow | |
| C8H7BrO | |
| 1 g | |
| CC1=C(C=C(C=C1)Br)C=O | |
| 199.047 | |
| 199.05 | |
| Crystalline Lumps |
| 47°C | |
| 172°C | |
| MFCD05662390 | |
| YILPAIKZHXATHY-UHFFFAOYSA-N | |
| 5-bromo-2-methylbenzaldehyde | |
| 10420289 | |
| ≥95.0% (GC) | |
| 5-Bromo-2-methylbenzaldehyde |
Safety and Handling
TSCA : No