missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylbenzaldehyde 95.0+%, TCI America™

Catalog No. B50365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50365G 5 g
B50361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50365G Supplier TCI America Supplier No. B50365G
Only null left

Chemical Identifiers

CAS 90050-59-2
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD05662390
InChI Key YILPAIKZHXATHY-UHFFFAOYSA-N
PubChem CID 10420289
IUPAC Name 5-bromo-2-methylbenzaldehyde
SMILES CC1=C(C=C(C=C1)Br)C=O

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 172°C
Quantity 5 g
Formula Weight 199.05
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Bromo-2-methylbenzaldehyde