missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-methylbenzaldehyde 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 47°C |
| Color | White-Yellow |
| Boiling Point | 172°C |
| Quantity | 1 g |
| Formula Weight | 199.05 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 5-Bromo-2-methylbenzaldehyde |
Safety and Handling
| TSCA | No |