missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylaniline 97.0+%, TCI America™

Catalog No. p-7123103
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36875G 5 g
B368725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36875G Supplier TCI America Supplier No. B36875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39478-78-9
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD00800678
InChI Key RXQNKKRGJJRMKD-UHFFFAOYSA-N
Synonym 2-amino-4-bromotoluene, 5-bromo-o-toluidine, 3-bromo-6-methylaniline, 5-bromo-2-methyl-aniline, benzenamine, 5-bromo-2-methyl, 5-bromo-2-methyl aniline, 5-bromo-2-methyl-phenylamine, zlchem 583, pubchem3801, acmc-1ajfs
PubChem CID 2734805
IUPAC Name 5-bromo-2-methylaniline
SMILES CC1=C(C=C(C=C1)Br)N

Specifications

Boiling Point 139°C
Quantity 5 g
Formula Weight 186.05
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Bromo-2-methylaniline