missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylaniline, 98%, Thermo Scientific Chemicals

Catalog No. p-3212214
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC372710250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC372710250 Supplier Thermo Scientific Chemicals Supplier No. 372710250

CAS: 39478-78-9 | Molecular Formula: C7H8BrN | Molecular Weight: 186.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39478-78-9
MDL Number MFCD00800678
InChI Key RXQNKKRGJJRMKD-UHFFFAOYSA-N
Synonym 2-amino-4-bromotoluene, 5-bromo-o-toluidine, 3-bromo-6-methylaniline, 5-bromo-2-methyl-aniline, benzenamine, 5-bromo-2-methyl, 5-bromo-2-methyl aniline, 5-bromo-2-methyl-phenylamine, zlchem 583, pubchem3801, acmc-1ajfs
PubChem CID 2734805
IUPAC Name 5-bromo-2-methylaniline
SMILES CC1=C(C=C(C=C1)Br)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.4900g/mL
Boiling Point 139°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8BrN
Refractive Index 1.6120 to 1.614
Linear Formula BrC6H3(CH3)NH2
Quantity 25 g
Specific Gravity 1.49
Molecular Weight (g/mol) 186.05
Formula Weight 186.05
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 5-Bromo-2-methylaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 254-467-2

RUO – Research Use Only