missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. B40701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40701G 1 g
B40705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40701G Supplier TCI America Supplier No. B40701G

Chemical Identifiers

CAS 911434-05-4
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD09031419
InChI Key FZZLWWNOYMHSIS-UHFFFAOYSA-N
Synonym 5-bromo-3-nitropicoline, 5-bromo-3-nitro-2-picoline, 5-bromo-2-methyl-3-nitro-pyridine, pyridine, 5-bromo-2-methyl-3-nitro, pubchem13809, ksc496e9t, bromo-2-methyl-3-nitropyridine
PubChem CID 25067266
IUPAC Name 5-bromo-2-methyl-3-nitropyridine
SMILES CC1=NC=C(C=C1[N+](=O)[O-])Br

Specifications

Melting Point 40°C
Color Yellow
Quantity 1 g
Formula Weight 217.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methyl-3-nitropyridine