missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methyl-2-pentene, 97%

Catalog No. AAL1940603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1940603 1 g
AAL1940606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1940603 Supplier Thermo Scientific Chemicals Supplier No. L1940603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2270-59-9 | C6H11Br | 163.06 g/mol

5-Bromo-2-methyl-2-pentene acts as a synthetic reagent, which is useful in the formation of sesqueterpenoid. It is also employed in the preparation of geranlol-3-14C. Further, it is used in the preparation of penifulvin A, which is a sesquiterpenoid with a novel dioxa-fenestrane structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-methyl-2-pentene acts as a synthetic reagent, which is useful in the formation of sesqueterpenoid. It is also employed in the preparation of geranlol-3-14C. Further, it is used in the preparation of penifulvin A, which is a sesquiterpenoid with a novel dioxa-fenestrane structure.

Solubility
Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 2270-59-9
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.06
MDL Number MFCD00009887
InChI Key UNXURIHDFUQNOC-UHFFFAOYSA-N
Synonym 5-bromo-2-methyl-2-pentene, acmc-20ah0m, 1-bromo4-methyl-3-pentene, 5-brom-2-methyl-2-pentene, 1-bromo-4-methyl-3-pentene, 2-methyl-5-bromo-2-pentene, 2-pentene,5-bromo-2-methyl, 5-bromo-2-methyl-pent-2-ene
PubChem CID 137521
IUPAC Name 5-bromo-2-methylpent-2-ene
SMILES CC(C)=CCCBr

Specifications

Density 1.219
Boiling Point 50°C (20 mmHg)
Flash Point 23°C (73°F)
Refractive Index 1.4775
Quantity 1 g
UN Number UN1993
Beilstein 1735749
Solubility Information Slightly miscible with water.
Formula Weight 163.06
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methyl-2-pentene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only