missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxytoluene 98.0+%, TCI America™

$103.97 - $404.28

Chemical Identifiers

CAS 14804-31-0
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD01321139
InChI Key UDLRGQOHGYWLCS-UHFFFAOYSA-N
Synonym 4-bromo-2-methylanisole, 5-bromo-2-methoxytoluene, benzene, 4-bromo-1-methoxy-2-methyl, 4-bromo-1-methoxy-2-methyl-benzene, 2-methyl-4-bromoanisole, 4-bromo2-methylanisole, acmc-1cfej, 4-bromo2-methyl-anisole, 4-bromo-2-methyl anisole, 4-bromo-2-methyl-anisole
PubChem CID 608315
IUPAC Name 4-bromo-1-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19105G
View Documents
TCI America
B19105G
5 g N/A
Only null left
 
B191025G
View Documents
TCI America
B191025G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

14804-31-0
201.063
UDLRGQOHGYWLCS-UHFFFAOYSA-N
608315
CC1=C(C=CC(=C1)Br)OC
C8H9BrO
MFCD01321139
4-bromo-2-methylanisole, 5-bromo-2-methoxytoluene, benzene, 4-bromo-1-methoxy-2-methyl, 4-bromo-1-methoxy-2-methyl-benzene, 2-methyl-4-bromoanisole, 4-bromo2-methylanisole, acmc-1cfej, 4-bromo2-methyl-anisole, 4-bromo-2-methyl anisole, 4-bromo-2-methyl-anisole
4-bromo-1-methoxy-2-methylbenzene

Specifications

14804-31-0
White-Yellow
C8H9BrO
5 g
UDLRGQOHGYWLCS-UHFFFAOYSA-N
4-bromo-1-methoxy-2-methylbenzene
608315
≥98.0% (GC)
5-Bromo-2-methoxytoluene
69°C
124°C
MFCD01321139
4-bromo-2-methylanisole, 5-bromo-2-methoxytoluene, benzene, 4-bromo-1-methoxy-2-methyl, 4-bromo-1-methoxy-2-methyl-benzene, 2-methyl-4-bromoanisole, 4-bromo2-methylanisole, acmc-1cfej, 4-bromo2-methyl-anisole, 4-bromo-2-methyl anisole, 4-bromo-2-methyl-anisole
CC1=C(C=CC(=C1)Br)OC
201.063
201.06
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'