missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxytoluene 98.0+%, TCI America™

Catalog No. B19105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19105G 5 g
B191025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19105G Supplier TCI America Supplier No. B19105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14804-31-0
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD01321139
InChI Key UDLRGQOHGYWLCS-UHFFFAOYSA-N
Synonym 4-bromo-2-methylanisole, 5-bromo-2-methoxytoluene, benzene, 4-bromo-1-methoxy-2-methyl, 4-bromo-1-methoxy-2-methyl-benzene, 2-methyl-4-bromoanisole, 4-bromo2-methylanisole, acmc-1cfej, 4-bromo2-methyl-anisole, 4-bromo-2-methyl anisole, 4-bromo-2-methyl-anisole
PubChem CID 608315
IUPAC Name 4-bromo-1-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)OC

Specifications

Melting Point 69°C
Color White-Yellow
Boiling Point 124°C
Quantity 5 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxytoluene