missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyridine, 98%

CAS: 13472-85-0 | C6H6BrNO | 188.024 g/mol

Supplier:  Thermo Scientific Chemicals L2001714

 View more versions of this product

Catalog No. AAL2001714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Bromo-2-methoxypyridine
1.453
96°C (204°F)
1.555
25 g
2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408
COC1=NC=C(C=C1)Br
188.024
188.02
13472-85-0
80°C (12 mmHg)
C6H6BrNO
MFCD01318952
115150
XADICJHFELMBGX-UHFFFAOYSA-N
5-bromo-2-methoxypyridine
2734895
98%

Chemical Identifiers

13472-85-0
188.024
XADICJHFELMBGX-UHFFFAOYSA-N
2734895
COC1=NC=C(C=C1)Br
C6H6BrNO
MFCD01318952
2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408
5-bromo-2-methoxypyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only