missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyridine, 98%

Catalog No. p-7047957
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2001706 5 g
AAL2001714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2001706 Supplier Thermo Scientific Chemicals Supplier No. L2001706
Only null left

CAS: 13472-85-0 | C6H6BrNO | 188.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13472-85-0
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD01318952
InChI Key XADICJHFELMBGX-UHFFFAOYSA-N
Synonym 2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408
PubChem CID 2734895
IUPAC Name 5-bromo-2-methoxypyridine
SMILES COC1=NC=C(C=C1)Br

Specifications

Density 1.453
Boiling Point 80°C (12 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.555
Quantity 5 g
Beilstein 115150
Formula Weight 188.02
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-methoxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only