missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-methoxyphenol 98.0+%, TCI America™
$38.51 - $98.18
Chemical Identifiers
| CAS | 37942-01-1 |
|---|---|
| Molecular Formula | C7H7BrO2 |
| Molecular Weight (g/mol) | 203.035 |
| MDL Number | MFCD00673137 |
| InChI Key | OLSJHVZRUFFIPL-UHFFFAOYSA-N |
| Synonym | 4-Bromo-2-hydroxyanisole |
| PubChem CID | 142228 |
| IUPAC Name | 5-bromo-2-methoxyphenol |
| SMILES | COC1=C(C=C(C=C1)Br)O |
Chemical Identifiers
| 37942-01-1 | |
| 203.035 | |
| OLSJHVZRUFFIPL-UHFFFAOYSA-N | |
| 142228 | |
| COC1=C(C=C(C=C1)Br)O |
| C7H7BrO2 | |
| MFCD00673137 | |
| 4-Bromo-2-hydroxyanisole | |
| 5-bromo-2-methoxyphenol |
Specifications
| 37942-01-1 | |
| White-Yellow | |
| C7H7BrO2 | |
| 5 g | |
| OLSJHVZRUFFIPL-UHFFFAOYSA-N | |
| 5-bromo-2-methoxyphenol | |
| 142228 | |
| ≥98.0% (GC,T) | |
| 5-Bromo-2-methoxyphenol |
| 65°C | |
| 150°C | |
| MFCD00673137 | |
| 4-Bromo-2-hydroxyanisole | |
| COC1=C(C=C(C=C1)Br)O | |
| 203.035 | |
| 203.04 | |
| Crystalline Powder |
Safety and Handling
TSCA : No