missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-methoxyphenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 65°C |
| Color | White-Yellow |
| Boiling Point | 150°C |
| Quantity | 5 g |
| Formula Weight | 203.04 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Bromo-2-methoxyphenol |
Safety and Handling
| TSCA | No |