missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. B47415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47415G 5 g
B47411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47415G Supplier TCI America Supplier No. B47415G
Only null left

Chemical Identifiers

CAS 2476-35-9
Molecular Formula C8H6BrO3
Molecular Weight (g/mol) 230.04
MDL Number MFCD00093713
InChI Key JFDUXZIRWBYBAQ-UHFFFAOYSA-M
Synonym 5-Bromo-o-anisic Acid
PubChem CID 881739
IUPAC Name 5-bromo-2-methoxybenzoate
SMILES COC1=CC=C(Br)C=C1C([O-])=O

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 5 g
Formula Weight 231.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxybenzoic Acid