missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxyaniline 97.0+%, TCI America™

Catalog No. p-7123186
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38281G 1 g
B38285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38281G Supplier TCI America Supplier No. B38281G

Chemical Identifiers

CAS 6358-77-6
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.05
MDL Number MFCD04037882
InChI Key OPGNSNDTPPIYPG-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxy aniline, 2-amino-4-bromoanisole, 5-bromo-o-anisidine, benzenamine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-aniline, 4-bromo-2-aminoanisole, 2-methoxy-5-bromoaniline, acmc-1b9k2, 5-bromo-2-methoxyphenylamine, 5-bromo-2-methyloxy aniline
PubChem CID 3585328
IUPAC Name 5-bromo-2-methoxyaniline
SMILES COC1=C(N)C=C(Br)C=C1

Specifications

Melting Point 95°C
Quantity 1 g
UN Number 2811
Formula Weight 202.05
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxyaniline