missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxyaniline, 97%

Catalog No. AAB2569306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2569306 5 g
AAB2569314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2569306 Supplier Thermo Scientific Chemicals Supplier No. B2569306

CAS: 6358-77-6 | C7H8BrNO | 202.05 g/mol

5-Bromo-2-methoxyaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-methoxyaniline is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Preserve in well- closed, light- resistant & tight container. Store in cool & dry place.

Chemical Identifiers

CAS 6358-77-6
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.05
MDL Number MFCD04037882
InChI Key OPGNSNDTPPIYPG-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxy aniline, 2-amino-4-bromoanisole, 5-bromo-o-anisidine, benzenamine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-aniline, 4-bromo-2-aminoanisole, 2-methoxy-5-bromoaniline, acmc-1b9k2, 5-bromo-2-methoxyphenylamine, 5-bromo-2-methyloxy aniline
PubChem CID 3585328
IUPAC Name 5-bromo-2-methoxyaniline
SMILES COC1=C(N)C=C(Br)C=C1

Specifications

Melting Point 94°C to 96°C
Quantity 5 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 202.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only