missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodobenzoic Acid 98.0+%, TCI America™

Catalog No. B42965G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B42965G Supplier TCI America Supplier No. B42965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21740-00-1
Molecular Formula C7H4BrIO2
Molecular Weight (g/mol) 326.915
MDL Number MFCD00079717
InChI Key IGBNDUKRHPTOBP-UHFFFAOYSA-N
Synonym 2-iodo-5-bromobenzoic acid, 5-bromo-2-iodo-benzoic acid, benzoic acid, 5-bromo-2-iodo, 5-bromo-2-iodobenzoicacid, pubchem3789, acmc-1bfz2, ksc497i6j, rarechem al bo 0892, akos bbv-001713, 5-bromo-2-iodobenzoic acid
PubChem CID 302939
IUPAC Name 5-bromo-2-iodobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)I

Specifications

Melting Point 162°C
Quantity 5 g
Formula Weight 326.92
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 5-Bromo-2-iodobenzoic Acid