missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodobenzoic acid, 97%

Catalog No. AAH6464306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6464306 5 g
AAH6464314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6464306 Supplier Thermo Scientific Chemicals Supplier No. H6464306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21740-00-1 | C7H4BrIO2 | 326.915 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.

Chemical Identifiers

CAS 21740-00-1
Molecular Formula C7H4BrIO2
Molecular Weight (g/mol) 326.915
MDL Number MFCD00079717
InChI Key IGBNDUKRHPTOBP-UHFFFAOYSA-N
Synonym 2-iodo-5-bromobenzoic acid, 5-bromo-2-iodo-benzoic acid, benzoic acid, 5-bromo-2-iodo, 5-bromo-2-iodobenzoicacid, pubchem3789, acmc-1bfz2, ksc497i6j, rarechem al bo 0892, akos bbv-001713, 5-bromo-2-iodobenzoic acid
PubChem CID 302939
IUPAC Name 5-bromo-2-iodobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)I

Specifications

Melting Point 158°C to 163°C
Quantity 5 g
Beilstein 3247380
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 326.92
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only