missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxypyrimidine 95.0+%, TCI America™

$61.51 - $208.78

Chemical Identifiers

CAS 38353-06-9
Molecular Formula C4H3BrN2O
Molecular Weight (g/mol) 174.985
InChI Key VTUDATOSQGYWML-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxypyrimidine, 5-bromopyrimidin-2-ol, 5-bromo-2-pyrimidinol, 5-bromo-2-pyrimidinone, 2 1h-pyrimidinone, 5-bromo, 5-bromopyrimidin-2 1h-one, 5-bromo-2 1h-pyrimidinone, 5-bromopyrimidin-2-one, 5-bromo-pyrimidin-2-ol, 2-pyrimidinol, 5-bromo
PubChem CID 101494
IUPAC Name 5-bromo-1H-pyrimidin-2-one
SMILES C1=C(C=NC(=O)N1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28485G
View Documents
TCI America
B28485G
5 g
Each for $61.51
Only null left
 
B284825G
View Documents
TCI America
B284825G
25 g
Each for $208.78
Only null left
 
Description

Description

Specifications

Chemical Identifiers

38353-06-9
174.985
5-bromo-2-hydroxypyrimidine, 5-bromopyrimidin-2-ol, 5-bromo-2-pyrimidinol, 5-bromo-2-pyrimidinone, 2 1h-pyrimidinone, 5-bromo, 5-bromopyrimidin-2 1h-one, 5-bromo-2 1h-pyrimidinone, 5-bromopyrimidin-2-one, 5-bromo-pyrimidin-2-ol, 2-pyrimidinol, 5-bromo
5-bromo-1H-pyrimidin-2-one
C4H3BrN2O
VTUDATOSQGYWML-UHFFFAOYSA-N
101494
C1=C(C=NC(=O)N1)Br

Specifications

38353-06-9
Yellow
5 g
VTUDATOSQGYWML-UHFFFAOYSA-N
5-bromo-1H-pyrimidin-2-one
101494
≥95.0% (GC)
5-Bromo-2-hydroxypyrimidine
235°C
C4H3BrN2O
5-bromo-2-hydroxypyrimidine, 5-bromopyrimidin-2-ol, 5-bromo-2-pyrimidinol, 5-bromo-2-pyrimidinone, 2 1h-pyrimidinone, 5-bromo, 5-bromopyrimidin-2 1h-one, 5-bromo-2 1h-pyrimidinone, 5-bromopyrimidin-2-one, 5-bromo-pyrimidin-2-ol, 2-pyrimidinol, 5-bromo
C1=C(C=NC(=O)N1)Br
174.985
174.99
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents