CAS RN 38353-06-9
IUPAC Name:
5-bromo-1,2-dihydropyrimidin-2-one
Synonyms:
5-bromo-1H-pyrimidin-2-one
Molecular Weight (g/mol):
174.99
Molecular Formula:
C4H3BrN2O
InChi Key:
VTUDATOSQGYWML-UHFFFAOYSA-N
SMILES:
BrC1=CNC(=O)N=C1
1
–
1
of
1
results
1
5-Bromo-2-hydroxypyrimidine 95.0+%, TCI America™
CAS: 38353-06-9 Molecular Formula: C4H3BrN2O Molecular Weight (g/mol): 174.985 InChI Key: VTUDATOSQGYWML-UHFFFAOYSA-N Synonym: 5-bromo-2-hydroxypyrimidine,5-bromopyrimidin-2-ol,5-bromo-2-pyrimidinol,5-bromo-2-pyrimidinone,2 1h-pyrimidinone, 5-bromo,5-bromopyrimidin-2 1h-one,5-bromo-2 1h-pyrimidinone,5-bromopyrimidin-2-one,5-bromo-pyrimidin-2-ol,2-pyrimidinol, 5-bromo PubChem CID: 101494 IUPAC Name: 5-bromo-1H-pyrimidin-2-one SMILES: C1=C(C=NC(=O)N1)Br