missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-hydroxypyridine 98.0+%, TCI America™
Supplier: TCI America B220725G
Specifications
| 5-Bromo-2-hydroxypyridine | |
| 180°C | |
| MFCD00234042 | |
| 5-bromo-2-hydroxypyridine, 5-bromopyridin-2-ol, 2-hydroxy-5-bromopyridine, 5-bromopyridin-2 1h-one, 5-bromo-2-pyridinol, 5-bromo-2 1h-pyridinone, 5-bromo-2 1h-pyridone, 2 1h-pyridinone, 5-bromo, 3-bromo-6-hydroxypyridine, 5-bromo-1,2-dihydropyridin-2-one | |
| C1=CC(=O)NC=C1Br | |
| 173.997 | |
| 174.00 | |
| Crystalline Powder |
| 13466-38-1 | |
| C5H4BrNO | |
| 25 g | |
| NDMZZQRNZFWMEZ-UHFFFAOYSA-N | |
| 5-bromo-1H-pyridin-2-one | |
| 599528 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 13466-38-1 | |
| 173.997 | |
| NDMZZQRNZFWMEZ-UHFFFAOYSA-N | |
| 599528 | |
| C1=CC(=O)NC=C1Br |
| C5H4BrNO | |
| MFCD00234042 | |
| 5-bromo-2-hydroxypyridine, 5-bromopyridin-2-ol, 2-hydroxy-5-bromopyridine, 5-bromopyridin-2 1h-one, 5-bromo-2-pyridinol, 5-bromo-2 1h-pyridinone, 5-bromo-2 1h-pyridone, 2 1h-pyridinone, 5-bromo, 3-bromo-6-hydroxypyridine, 5-bromo-1,2-dihydropyridin-2-one | |
| 5-bromo-1H-pyridin-2-one |
Safety and Handling
TSCA : No