missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxypyridine 98.0+%, TCI America™

Catalog No. p-7119554
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22075G 5 g
B220725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22075G Supplier TCI America Supplier No. B22075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13466-38-1 | C5H4BrNO | 173.997 g/mol

Chemical Identifiers

CAS 13466-38-1
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 173.997
MDL Number MFCD00234042
InChI Key NDMZZQRNZFWMEZ-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxypyridine, 5-bromopyridin-2-ol, 2-hydroxy-5-bromopyridine, 5-bromopyridin-2 1h-one, 5-bromo-2-pyridinol, 5-bromo-2 1h-pyridinone, 5-bromo-2 1h-pyridone, 2 1h-pyridinone, 5-bromo, 3-bromo-6-hydroxypyridine, 5-bromo-1,2-dihydropyridin-2-one
PubChem CID 599528
IUPAC Name 5-bromo-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1Br

Specifications

Melting Point 180°C
Quantity 5 g
Formula Weight 174.00
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydroxypyridine