missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-hydroxybenzyl Alcohol 98.0+%, TCI America™
$44.73 - $128.62
Chemical Identifiers
| CAS | 2316-64-5 |
|---|---|
| Molecular Formula | C7H7BrO2 |
| Molecular Weight (g/mol) | 203.04 |
| MDL Number | MFCD00004618 |
| InChI Key | KNKRHSVKIORZQB-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e |
| PubChem CID | 75342 |
| IUPAC Name | 4-bromo-2-(hydroxymethyl)phenol |
| SMILES | OCC1=CC(Br)=CC=C1O |
Chemical Identifiers
| 2316-64-5 | |
| 203.04 | |
| KNKRHSVKIORZQB-UHFFFAOYSA-N | |
| 75342 | |
| OCC1=CC(Br)=CC=C1O |
| C7H7BrO2 | |
| MFCD00004618 | |
| 5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e | |
| 4-bromo-2-(hydroxymethyl)phenol |
Specifications
| 2316-64-5 | |
| White-Yellow | |
| MFCD00004618 | |
| 5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e | |
| OCC1=CC(Br)=CC=C1O | |
| 203.04 | |
| 203.04 | |
| Crystalline Powder |
| 108°C | |
| C7H7BrO2 | |
| 1 g | |
| KNKRHSVKIORZQB-UHFFFAOYSA-N | |
| 4-bromo-2-(hydroxymethyl)phenol | |
| 75342 | |
| ≥98.0% (GC) | |
| 5-Bromo-2-hydroxybenzyl Alcohol |
Safety and Handling
TSCA : No