missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. B21641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21641G 1 g
B21645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21641G Supplier TCI America Supplier No. B21641G

Chemical Identifiers

CAS 2316-64-5
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.04
MDL Number MFCD00004618
InChI Key KNKRHSVKIORZQB-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e
PubChem CID 75342
IUPAC Name 4-bromo-2-(hydroxymethyl)phenol
SMILES OCC1=CC(Br)=CC=C1O

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 1 g
Formula Weight 203.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydroxybenzyl Alcohol